1-(2-Chloro-6-fluorobenzyl)piperazin

650156-10G 650156-1G
220085-119EA 41.31 USD
220085-119 220085-120
1-(2-Chloro-6-fluorobenzyl)piperazin
1-(2-Chloro-6-fluorobenzyl)piperazin
Formula: C₁₁H₁₄ClFN₂
MW: 228.7 g/mol
MDL Number: MFCD00816605
CAS Number: 215655-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR