2-Bromo-3-methoxypyridine

649716-10G 649716-1G
220085-084EA 299.38 USD
220085-084 220085-085
2-Bromo-3-methoxypyridine
2-Bromo-3-methoxypyridine
Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD01570896
CAS Number: 24100-18-3
EINECS: 246-017-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR