6-Chloroquinaldine ≥97%

649260-25G 649260-5G
220085-049EA 317.69 USD
220085-049 220084-376
6-Chloroquinaldine ≥97%
6-Chloroquinaldine
Formula: C₁₀H₈ClN
MW: 177.63 g/mol
MDL Number: MFCD00051735
CAS Number: 92-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR