2,6-Dichloro-3-(trifluoromethyl)pyridine

648884-1G 648884-250MG
220085-018EA 195 USD
220085-018 220085-019
2,6-Dichloro-3-(trifluoromethyl)pyridine
2,6-Dichloro-3-(trifluoromethyl)pyridine
Formula: C₆H₂Cl₂F₃N
MW: 215.99 g/mol
Boiling Pt: 195 °C (1013 hPa)
MDL Number: MFCD00042245
CAS Number: 55304-75-1
EINECS: 259-585-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR