3'-Chloro-4'-methoxyacetophenone ≥99%

Synonyms: 1-(3-Chloro-4-methoxyphenyl)ethan-1-one, 1-(3-Chloro-4-methoxyphenyl)ethanone, 1-(3-Chloro-4-methoxyphenyl)ethanone

648809-1G 648809-5G
220085-012EA 40.92 USD
220085-012 220085-013
3'-Chloro-4'-methoxyacetophenone ≥99%
3'-Chloro-4'-methoxyacetophenone
Formula: C₉H₉ClO₂
MW: 184.62 g/mol
MDL Number: MFCD04973413
CAS Number: 37612-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR