4-Bromo-2-methylthiophene

648728-1G 648728-5G
220085-004EA 89.77 USD
220085-004 220085-005
4-Bromo-2-methylthiophene
4-Bromo-2-methylthiophene
Formula: C₅H₅BrS
MW: 177.06 g/mol
MDL Number: MFCD04116367
CAS Number: 29421-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR