5-Iodo-o-toluidine ≥97%

Synonyms: 2-Amino-4-iodotoluene, 5-Iodo-2-methylaniline

648647-1G 648647-5G
220084-997EA 72.15 USD
220084-997 220084-998
5-Iodo-o-toluidine ≥97%
5-Iodo-o-toluidine
Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 48 - 49 °C
MDL Number: MFCD00051531
CAS Number: 83863-33-6
EINECS: 281-094-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR