1-Methyl-3-phenylpiperazine

Danger

Synonyms: MEFPIP

648434-5G 648434-1G
220084-755EA 180.92 USD
220084-755 220084-754
1-Methyl-3-phenylpiperazine
1-Methyl-3-phenylpiperazine
Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD03411603
CAS Number: 5271-27-2
EINECS: 431-180-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR