2-Amino-5-bromobenzothiazole

647683-1G 647683-5G
220084-927EA 621.69 USD
220084-927 220084-928
2-Amino-5-bromobenzothiazole
2-Amino-5-bromobenzothiazole
Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
MDL Number: MFCD00577806
CAS Number: 20358-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR