Methyl-4-bromo-2-methoxybenzoate ≥98%

Synonyms: Methyl 4-bromo-2-methoxybenzoate, 4-Bromo-2-methoxybenzoic acid methyl ester

647594-1G 647594-10G
220084-911EA 38.77 USD
220084-911 220084-910
Methyl-4-bromo-2-methoxybenzoate ≥98%
Methyl-4-bromo-2-methoxybenzoate
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD06797867
CAS Number: 139102-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR