3-Chloro-2-nitrobenzyl alcohol ≥97%

647586-5G 647586-1G
220084-908EA 185.23 USD
220084-908 220084-907
3-Chloro-2-nitrobenzyl alcohol ≥97%
3-Chloro-2-nitrobenzyl alcohol
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04038814
CAS Number: 77158-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR