4-Chloro-2-methylbenzyl alcohol ≥97%

647551-1G 647551-5G
220084-902EA 59.23 USD
220084-902 220084-903
4-Chloro-2-methylbenzyl alcohol ≥97%
4-Chloro-2-methylbenzyl alcohol
Formula: C₈H₉ClO
MW: 156.61 g/mol
MDL Number: MFCD06201140
CAS Number: 129716-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR