Silicon

647543-1EA 647543-5EA
220084-900EA 363.54 USD
220084-900 220084-901
Silicon
Silicon

Silicon Wafer (Single Side Polished) (111) Undoped (N-Type) Diam. X Thickness 3 In. X 0.5 mm

Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR