2-Chloro-4-iodopyridine

647403-5G 647403-1G
220084-886EA 266 USD
220084-886 220084-885
2-Chloro-4-iodopyridine
2-Chloro-4-iodopyridine
Formula: C₅H₃ClIN
MW: 239.44 g/mol
MDL Number: MFCD01861983
CAS Number: 153034-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR