2-Chloro-3-hydroxybenzaldehyde ≥97%

647365-1G 647365-10G
220084-881EA 62.14 USD
220084-881 220084-880
2-Chloro-3-hydroxybenzaldehyde ≥97%
2-Chloro-3-hydroxybenzaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
MDL Number: MFCD06798118
CAS Number: 56962-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR