6-Bromonicotinic acid

Synonyms: 6-Bromopyridine-3-carboxylic acid, 2-Bromo-5-pyridinecarboxylic acid, 6-Bromo-3-pyridinecarboxylic acid

646989-10G 646989-1G
220084-816EA 215.38 USD
220084-816 220084-817
6-Bromonicotinic acid
6-Bromonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
MDL Number: MFCD01927100
CAS Number: 6311-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR