4-Amino-3-nitropyridine

Warning

Synonyms: 3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine

646962-1G 646962-10G
220084-814EA 42.43 USD
220084-814 220084-813
4-Amino-3-nitropyridine
4-Amino-3-nitropyridine
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
MDL Number: MFCD00160304
CAS Number: 1681-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR