(3-Iodophenyl)acetonitrile ≥90%

Synonyms: 3-Iodobenzyl cyanide, 2-(3-Iodophenyl)acetonitrile

646415-5G 646415-1G
220084-715EA 213.23 USD
220084-715 220084-714
(3-Iodophenyl)acetonitrile ≥90%
(3-Iodophenyl)acetonitrile
Formula: C₈H₆IN
MW: 243.05 g/mol
Boiling Pt: 138 °C (12 mmHg)
Melting Pt: 33…35 °C
MDL Number: MFCD00040890
CAS Number: 130723-54-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR