4-Fluoro-3-iodotoluene ≥97%

646334-1G 646334-5G
220084-689EA 491.69 USD
220084-689 220084-690
4-Fluoro-3-iodotoluene ≥97%
4-Fluoro-3-iodotoluene
Formula: C₇H₆FI
MDL Number: MFCD00079759
CAS Number: 452-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR