5-Bromo-2-ethoxypyridine

646326-5G 646326-1G
220084-688EA 287.54 USD
220084-688 220084-687
5-Bromo-2-ethoxypyridine
5-Bromo-2-ethoxypyridine
Formula: C₇H₈BrNO
MW: 202.05 g/mol
MDL Number: MFCD00234311
CAS Number: 55849-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR