4-Bromo-2-chloropyridine

646318-25G 646318-5G
220084-684EA 491.69 USD
220084-684 220084-685
4-Bromo-2-chloropyridine
4-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
MDL Number: MFCD03840756
CAS Number: 73583-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR