3-Bromo-5-fluoropyridine

646296-1G 646296-5G
220084-679EA 30.15 USD
220084-679 220084-680
3-Bromo-5-fluoropyridine
3-Bromo-5-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
MDL Number: MFCD04112555
CAS Number: 407-20-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR