1-(4-Fluorobenzyl)piperazin

646164-5G 646164-1G
220082-416EA 105.65 USD
220082-416 220082-415
1-(4-Fluorobenzyl)piperazin
1-(4-Fluorobenzyl)piperazin
Formula: C₁₁H₁₅FN₂
MW: 194.25 g/mol
MDL Number: MFCD01075234
CAS Number: 70931-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR