Indole-4-carbonitrile

Warning

Synonyms: 4-Cyano-1H-indole, 1H-Indole-4-carbonitrile, 4-Cyanoindole

645532-5G 645532-1G
220084-539EA 165.85 USD
220084-539 220084-538
Indole-4-carbonitrile
Indole-4-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
MDL Number: MFCD00152045
CAS Number: 16136-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR