5-Amino-3-phenylisoxazole

644978-5G 644978-1G
220084-506EA 258.46 USD
220084-506 220084-505
5-Amino-3-phenylisoxazole
5-Amino-3-phenylisoxazole
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00464114
CAS Number: 4369-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR