3-Chloro-4-hydroxybenzaldehyde ≥97%

644188-1G 644188-5G
220084-444EA 41.25 USD
220084-444 220084-445
3-Chloro-4-hydroxybenzaldehyde ≥97%
3-Chloro-4-hydroxybenzaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 133 - 134 °C
Density: 1.0404 g/cm³ (20 °C)
MDL Number: MFCD00016981
CAS Number: 2420-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR