2-(Trifluoromethyl)pyridine

643572-1G 643572-5G
220083-028EA 23.69 USD
220083-028 220083-249
2-(Trifluoromethyl)pyridine
2-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 138…139 °C (1013 hPa)
MDL Number: MFCD02183562
CAS Number: 368-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR