2-Chloro-5-fluoropyridine

643556-1G 643556-5G
220083-025EA 26.92 USD
220083-025 220083-026
2-Chloro-5-fluoropyridine
2-Chloro-5-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
MDL Number: MFCD03095332
CAS Number: 31301-51-6
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR