2-Bromo-6-nitrophenol ≥97%

643378-5G 643378-25G
220084-352EA 113.08 USD
220084-352 220084-351
2-Bromo-6-nitrophenol ≥97%
2-Bromo-6-nitrophenol
Formula: C₆H₄BrNO₃
MDL Number: MFCD02683216
CAS Number: 13073-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR