2-Amino-5-fluorobenzonitrile ≥97%

642924-1G 642924-10G
220083-576EA 37.69 USD
220083-576 220083-575
2-Amino-5-fluorobenzonitrile ≥97%
2-Amino-5-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
MDL Number: MFCD02683869
CAS Number: 61272-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR