2-Amino-6-chlorobenzonitrile ≥96%

Warning

642843-1G 642843-5G
220084-197EA 50.83 USD
220084-197 220084-198
2-Amino-6-chlorobenzonitrile ≥96%
2-Amino-6-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 136 - 138 °C
MDL Number: MFCD00015546
CAS Number: 6575-11-7
EINECS: 229-501-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR