2-Amino-3,5-dichlorobenzonitrile

642819-1G 642819-5G
220084-191EA 41.78 USD
220084-191 220084-192
2-Amino-3,5-dichlorobenzonitrile
2-Amino-3,5-dichlorobenzonitrile
Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD00158947
CAS Number: 36764-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR