4-Amino-3,5-dichlorobenzonitrile ≥97%

642797-1G 642797-5G
220084-185EA 31.23 USD
220084-185 220084-186
4-Amino-3,5-dichlorobenzonitrile ≥97%
4-Amino-3,5-dichlorobenzonitrile
Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 120 °C
MDL Number: MFCD00017341
CAS Number: 78473-00-4
EINECS: 278-914-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR