3-Hydroxycoumarin

Synonyms: 3-Hydroxy-2-benzopyrone, 3-Hydroxy-2H-chromen-2-one

642673-1G 642673-5G
220084-170EA 128.15 USD
220084-170 220084-171
3-Hydroxycoumarin
3-Hydroxycoumarin
Formula: C₉H₆O₃
MW: 162.14 g/mol
MDL Number: MFCD00017490
CAS Number: 939-19-5
EINECS: 213-355-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR