6-Hydroxycoumarin

Synonyms: 6-Hydroxy-2-benzopyrone, 6-Hydroxy-2H-chromen-2-one

642665-1G 642665-5G
220084-168EA 74.2 USD
220084-168 220084-169
6-Hydroxycoumarin
6-Hydroxycoumarin
Formula: C₉H₆O₃
MW: 162.14 g/mol
MDL Number: MFCD00630040
CAS Number: 6093-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR