5-Bromothiazole

642517-5G 642517-1G
220084-148EA 296.15 USD
220084-148 220084-147
5-Bromothiazole
5-Bromothiazole
Formula: C₃H₂BrNS
MDL Number: MFCD07787394
CAS Number: 3034-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR