2-Bromo-5-thiazolecarboxylic acid

642487-5G 642487-1G
220084-141EA 122.15 USD
220084-141 220084-140
2-Bromo-5-thiazolecarboxylic acid
2-Bromo-5-thiazolecarboxylic acid
Formula: C₄H₂BrNO₂S
MDL Number: MFCD04115730
CAS Number: 54045-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR