Cadmium(II) nitrate tetrahydrate

Warning

Synonyms: Cadmium nitrate tetrahydrate

642045-500G 642045-100G
220084-100EA 220 USD
220084-100 220084-099
Cadmium(II) nitrate tetrahydrate
Cadmium(II) nitrate tetrahydrate
Formula: CdN₂O₆·4H₂O
MW: 308.48 g/mol
Boiling Pt: 132 °C (1013 hPa)
Melting Pt: 60 °C
Density: 2.5 g/cm³ (20 °C)
MDL Number: MFCD00149626
CAS Number: 10022-68-1
EINECS: 233-710-6
UN: 3087
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR