Ethyl-4-iodobenzoate ≥97%

Synonyms: Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester

641979-5G 641979-25G
220084-094EA 52.55 USD
220084-094 220084-093
Ethyl-4-iodobenzoate ≥97%
Ethyl-4-iodobenzoate
Formula: C₉H₉IO₂
MW: 276.07 g/mol
Boiling Pt: 281 °C (1013 hPa)
Density: 1.655 g/cm³ (20 °C)
MDL Number: MFCD00017344
CAS Number: 51934-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR