1-(3-Hydroxypropyl)piperazine

Synonyms: 1-Piperazinepropanol, 3-Piperazin-1-ylpropanol, 3-(Piperazin-1-yl)propan-1-ol

641642-1G 641642-5G
220084-075EA 102.8 USD
220084-075 220084-076
1-(3-Hydroxypropyl)piperazine
1-(3-Hydroxypropyl)piperazine
Formula: C₇H₁₆N₂O
MW: 144.22 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 49 °C
MDL Number: MFCD00023132
CAS Number: 5317-32-8
EINECS: 226-176-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR