2-Chloro-4,6-difluoroaniline ≥97%

641413-5G 641413-1G
220084-056EA 98.75 USD
220084-056 220084-055
2-Chloro-4,6-difluoroaniline ≥97%
2-Chloro-4,6-difluoroaniline
Formula: C₆H₄ClF₂N
MW: 163.55 g/mol
MDL Number: MFCD00142892
CAS Number: 36556-56-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR