1-Butyl-3-methylimidazolium bromide

64133-5G 64133-25G
220084-046EA 188 USD
220084-046 220084-045
1-Butyl-3-methylimidazolium bromide
1-Butyl-3-methylimidazolium bromide
Formula: C₈H₁₅BrN₂
MDL Number: MFCD03427611
CAS Number: 85100-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR