5-Amino-2-(trifluoromethyl)pyridine

Synonyms: 3-Amino-6-(trifluoromethyl)pyridine, 2-(Trifluoromethyl)pyridin-5-amine, 2-(Trifluoromethyl)-5-pyridylamine

640107-1G 640107-5G
220083-555EA 90.46 USD
220083-555 220083-556
5-Amino-2-(trifluoromethyl)pyridine
5-Amino-2-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
MDL Number: MFCD01862656
CAS Number: 106877-33-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR