5-Amino-2-fluorobenzonitrile ≥97%

Synonyms: 3-Cyano-4-fluoroaniline

639877-5G 639877-25G
220083-962EA 81.63 USD
220083-962 220083-961
5-Amino-2-fluorobenzonitrile ≥97%
5-Amino-2-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
MDL Number: MFCD00277872
CAS Number: 53312-81-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR