4-Phenylpiperidine

639869-1G 639869-5G
220083-959EA 58.58 USD
220083-959 220083-960
4-Phenylpiperidine
4-Phenylpiperidine
Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: 58…62 °C
Density: 0.967 g/cm³ (20 °C)
MDL Number: MFCD00006002
CAS Number: 771-99-3
EINECS: 212-243-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR