2-Bromo-1-methylimidazole

Synonyms: 2-Bromo-1-methyl-1H-imidazole

639850-1G 639850-5G
220083-957EA 131.38 USD
220083-957 220083-958
2-Bromo-1-methylimidazole
2-Bromo-1-methylimidazole
Formula: C₄H₅BrN₂
MW: 161 g/mol
MDL Number: MFCD02179525
CAS Number: 16681-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR