2-Fluoro-5-pyridineboronic acid

Synonyms: 6-Fluoropyridine-3-boronic acid, 2-Fluoropyridine-5-boronic acid, 2-Fluoro-5-pyridylboronic acid, 2-Fluoro-5-pyridinylboronic acid, 6-Fluoro-3-pyridinylboronic acid

639184-1G 639184-5G
220083-890EA 17.31 USD
220083-890 220083-891
2-Fluoro-5-pyridineboronic acid
2-Fluoro-5-pyridineboronic acid
Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
MDL Number: MFCD03411559
CAS Number: 351019-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR