(4-Chlorophenyl)acetyl chloride ≥97%

Danger

Synonyms: 2-(4-Chlorophenyl)acetyl chloride, 4-Chlorobenzeneacetyl chloride

638951-25G 638951-5G
220083-862EA 185.77 USD
220083-862 220083-863
(4-Chlorophenyl)acetyl chloride ≥97%
(4-Chlorophenyl)acetyl chloride
Formula: (C₆H₄Cl)CH₂COCl
MW: 189.04 g/mol
Boiling Pt: 85 °C (1 -rr)
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00037111
CAS Number: 25026-34-0
EINECS: 246-571-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR