2-Chloro-4-cyanobenzenesulfonyl chloride ≥97%

Synonyms: 2-Chloro-4-cyanobenzene-1-sulphonyl chloride

638870-1G 638870-5G
220083-850EA 116.31 USD
220083-850 220083-851
2-Chloro-4-cyanobenzenesulfonyl chloride ≥97%
2-Chloro-4-cyanobenzenesulphonyl chloride
Formula: C₇H₃Cl₂NO₂S
MW: 236.08 g/mol
MDL Number: MFCD00177089
CAS Number: 254749-11-6
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR