7-Nitroindole

638854-5G 638854-1G
220083-847EA 248.77 USD
220083-847 220083-846
7-Nitroindole
7-Nitroindole
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00005683
CAS Number: 6960-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR